Assay Details
| Gene Symbol: CCL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015438190.1 | XP_015293676.1 | 1 - 2 | 341 | 101 | |
| XM_015438191.1 | XP_015293677.1 | 3 - 4 | 244 | 101 | ||
More Information
Set Membership: |
|

CCL14
Genomic Map