Assay Details
| Gene Symbol: MFSD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551600.2 | XP_005551657.2 | 2 - 3 | 616 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MFSD4B| Gene Symbol: MFSD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551600.2 | XP_005551657.2 | 2 - 3 | 616 | 110 | |
Set Membership: |
|