Assay Details
Gene Symbol: SH3BGRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552497.2 | XP_005552554.1 | 3 - 4 | 1059 | 107 | |
XM_005552498.2 | XP_005552555.1 | 3 - 4 | 669 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
