Assay Details
| Gene Symbol: C4H6orf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449327.1 | XP_015304813.1 | 1 - 2 | 2172 | 81 | |
| XM_015449328.1 | XP_015304814.1 | 2 - 3 | 1378 | 81 | ||
More Information
Set Membership: |
|

C4H6orf141
Genomic Map