Assay Details
Gene Symbol: PCDHB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558029.2 | XP_005558086.1 | 1 | 1461 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
