Assay Details
Gene Symbol: LRRC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005561855.2 | XP_005561912.1 | 4 - 5 | 809 | 78 | |
XM_005561856.2 | XP_005561913.1 | 4 - 5 | 658 | 78 | ||
XM_005561857.2 | XP_005561914.1 | 4 - 5 | 809 | 78 | ||
XM_005561858.2 | XP_005561915.1 | 4 - 5 | 770 | 78 | ||
XM_015454028.1 | XP_015309514.1 | 4 - 5 | 809 | 78 | ||
XM_015454029.1 | XP_015309515.1 | 4 - 5 | 657 | 78 | ||
XM_015454030.1 | XP_015309516.1 | 4 - 5 | 809 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
