Assay Details
| Gene Symbol: DUSP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565420.2 | XP_005565477.1 | 3 - 4 | 496 | 75 | |
| XM_005565421.2 | XP_005565478.1 | 2 - 3 | 366 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUSP29| Gene Symbol: DUSP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565420.2 | XP_005565477.1 | 3 - 4 | 496 | 75 | |
| XM_005565421.2 | XP_005565478.1 | 2 - 3 | 366 | 75 | ||