Assay Details
Gene Symbol: APOBEC3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567248.2 | XP_005567305.1 | 2 - 3 | 433 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
