Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568475.2 | XP_005568532.1 | 3 - 4 | 1692 | 94 | |
| XM_005568476.2 | XP_005568533.1 | 3 - 4 | 1754 | 94 | ||
| XM_005568477.2 | XP_005568534.1 | 3 - 4 | 1714 | 94 | ||
| XM_015457838.1 | XP_015313324.1 | 2 - 3 | 2165 | 94 | ||
| XM_015457839.1 | XP_015313325.1 | 2 - 3 | 1529 | 94 | ||
More Information
Set Membership: |
|

LZTS3
Genomic Map