Assay Details
Gene Symbol: CAPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015431216.1 | XP_015286702.1 | 13 - 14 | 1508 | 89 | |
XM_015431217.1 | XP_015286703.1 | 13 - 14 | 1504 | 89 | ||
XM_015431218.1 | XP_015286704.1 | 13 - 14 | 1386 | 89 | ||
XM_015431219.1 | XP_015286705.1 | 13 - 14 | 1211 | 89 | ||
XM_015431220.1 | XP_015286706.1 | 12 - 13 | 1434 | 89 | ||
XM_015431221.1 | XP_015286707.1 | 12 - 13 | 1150 | 89 | ||
XM_015431222.1 | XP_015286708.1 | 11 - 12 | 1111 | 89 | ||
XM_015431223.1 | XP_015286709.1 | 8 - 9 | 817 | 89 | ||
XM_015431224.1 | XP_015286710.1 | 7 - 8 | 800 | 89 | ||
XM_015431225.1 | XP_015286711.1 | 8 - 9 | 762 | 89 | ||
XM_015431226.1 | XP_015286712.1 | 8 - 9 | 726 | 89 | ||
XM_015431227.1 | XP_015286713.1 | 3 - 4 | 1963 | 89 | ||
XM_015431228.1 | XP_015286714.1 | 6 - 7 | 714 | 89 | ||
GenBank mRNA | AB052179.1 | - | 5 - 6 | 443 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
