Assay Details
| Gene Symbol: IGFBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581246.2 | XP_005581303.1 | 3 - 4 | 746 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IGFBPL1| Gene Symbol: IGFBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581246.2 | XP_005581303.1 | 3 - 4 | 746 | 68 | |
Set Membership: |
|