Assay Details
| Gene Symbol: TLCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582447.2 | XP_005582504.1 | 1 - 2 | 6007 | 69 | |
| XM_015437429.1 | XP_015292915.1 | 4 - 5 | 4689 | 69 | ||
| XM_015437430.1 | XP_015292916.1 | 2 - 3 | 5650 | 69 | ||
| XM_015437431.1 | XP_015292917.1 | 3 - 4 | 5053 | 69 | ||
| XM_015437432.1 | XP_015292918.1 | 3 - 4 | 4608 | 69 | ||
| XM_015437433.1 | XP_015292919.1 | 5 - 6 | 4340 | 69 | ||
| XM_015437434.1 | XP_015292920.1 | 5 - 6 | 3926 | 69 | ||
| XM_015437435.1 | XP_015292921.1 | 5 - 6 | 3781 | 69 | ||
More Information
Set Membership: |
|

TLCD2
Genomic Map