Assay Details
Gene Symbol: KASH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589892.2 | XP_005589949.1 | 8 - 9 | 947 | 65 | |
XM_005589893.2 | XP_005589950.1 | 7 - 8 | 824 | 65 | ||
XM_005589894.2 | XP_005589951.1 | 7 - 8 | 824 | 65 | ||
XM_015441906.1 | XP_015297392.1 | 8 - 9 | 947 | 65 | ||
XM_015441908.1 | XP_015297394.1 | 8 - 9 | 947 | 65 | ||
XM_015441909.1 | XP_015297395.1 | 9 - 10 | 1002 | 65 | ||
XM_015441910.1 | XP_015297396.1 | 6 - 7 | 1130 | 65 | ||
XM_015441911.1 | XP_015297397.1 | 8 - 9 | 866 | 65 | ||
XM_015441912.1 | XP_015297398.1 | 7 - 8 | 1804 | 65 | ||
XM_015441913.1 | XP_015297399.1 | 8 - 9 | 2080 | 65 | ||
XM_015441914.1 | XP_015297400.1 | 7 - 8 | 1944 | 65 | ||
XM_015441915.1 | XP_015297401.1 | 6 - 7 | 1840 | 65 | ||
XM_015441916.1 | XP_015297402.1 | 6 - 7 | 1749 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
