Assay Details
Gene Symbol: LOC102125710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441930.1 | XP_015297416.1 | 3 - 4 | 838 | 108 |
Gene Symbol: LOC102125335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015440973.1 | XP_015296459.1 | 4 - 5 | 711 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
