Assay Details
Gene Symbol: LOC102115705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546925.2 | XP_005546982.1 | 3 - 4 | 772 | 78 | |
XM_005546926.2 | XP_005546983.1 | 3 - 4 | 776 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
