Assay Details
| Gene Symbol: LOC102141127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319498.1 | NP_001306427.1 | 1 - 2 | 308 | 78 | |
| GenBank mRNA | AB059654.1 | - | 1 - 2 | 308 | 78 | |
More Information
Set Membership: |
|

LOC102141127
Genomic Map