Assay Details
| Gene Symbol: MUC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549281.2 | XP_005549338.1 | 14714 | 78 | ||
| XM_015447181.1 | XP_015302667.1 | 14715 | 78 | |||
| XM_015447183.1 | XP_015302669.1 | 3 - 4 | 2879 | 78 | ||
More Information
Set Membership: |
|

MUC3A
Genomic Map