Assay Details
| Gene Symbol: GNRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555188.2 | XP_005555245.1 | 2 - 3 | 2485 | 75 | |
| XM_005555189.2 | XP_005555246.1 | 2 - 3 | 2357 | 75 | ||
More Information
Set Membership: |
|

GNRHR
Genomic Map