Assay Details
| Gene Symbol: ENC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557146.2 | XP_005557203.1 | 2 - 3 | 2990 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENC1| Gene Symbol: ENC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557146.2 | XP_005557203.1 | 2 - 3 | 2990 | 102 | |
Set Membership: |
|