Assay Details
| Gene Symbol: POMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283991.1 | NP_001270920.1 | 4 - 5 | 516 | 78 | |
| XM_015454372.1 | XP_015309858.1 | 3 - 4 | 984 | 78 | ||
| XM_015454373.1 | XP_015309859.1 | 4 - 5 | 626 | 78 | ||
| XM_015454374.1 | XP_015309860.1 | 3 - 4 | 495 | 78 | ||
| GenBank mRNA | AB070191.1 | - | 4 - 5 | 516 | 78 | |
More Information
Set Membership: |
|

POMK
Genomic Map