Assay Details
Gene Symbol: APCDD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015457702.1 | XP_015313188.1 | 1 - 2 | 3468 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: APCDD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015457702.1 | XP_015313188.1 | 1 - 2 | 3468 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() |