Assay Details
Gene Symbol: RFX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575105.2 | XP_005575162.1 | 10 - 11 | 975 | 81 | |
XM_005575107.2 | XP_005575164.1 | 10 - 11 | 1005 | 81 | ||
XM_005575108.2 | XP_005575165.1 | 10 - 11 | 1264 | 81 | ||
XM_005575111.2 | XP_005575168.1 | 8 - 9 | 761 | 81 | ||
XM_005575112.2 | XP_005575169.1 | 8 - 9 | 856 | 81 | ||
XM_005575113.2 | XP_005575170.1 | 8 - 9 | 1113 | 81 | ||
XM_015433281.1 | XP_015288767.1 | 9 - 10 | 896 | 81 | ||
XM_015433282.1 | XP_015288768.1 | 10 - 11 | 980 | 81 | ||
XM_015433283.1 | XP_015288769.1 | 10 - 11 | 985 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
