Assay Details
Gene Symbol: LOC102120532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579495.2 | XP_005579552.1 | 7 - 8 | 1261 | 74 | |
XM_015435645.1 | XP_015291131.1 | 6 - 7 | 1792 | 74 | ||
XM_015435646.1 | XP_015291132.1 | 6 - 7 | 1790 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
