Assay Details
| Gene Symbol: LOC102138104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589743.2 | XP_005589800.1 | 4 - 5 | 422 | 97 | |
| XM_005589744.2 | XP_005589801.1 | 3 - 4 | 242 | 97 | ||
| XM_015441323.1 | XP_015296809.1 | 3 - 4 | 583 | 97 | ||
| XM_015441324.1 | XP_015296810.1 | 3 - 4 | 256 | 97 | ||
More Information
Set Membership: |
|

LOC102138104
Genomic Map