Assay Details
Gene Symbol: LOC102143618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015445457.1 | XP_015300943.1 | 15 - 16 | 1435 | 95 |
Gene Symbol: LOC102142866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015444358.1 | XP_015299844.1 | 1588 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
