Assay Details
| Gene Symbol: NSMCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564053.2 | XP_005564110.1 | 5 - 6 | 658 | 69 | |
| XM_005564054.2 | XP_005564111.1 | 6 - 7 | 764 | 69 | ||
| XM_005564055.2 | XP_005564112.1 | 5 - 6 | 619 | 69 | ||
| XM_015455167.1 | XP_015310653.1 | 6 - 7 | 683 | 69 | ||
More Information
Set Membership: |
|

NSMCE2
Genomic Map