Assay Details
Gene Symbol: LOC102125578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567341.2 | XP_005567398.1 | 4 - 5 | 884 | 93 | |
XM_015457223.1 | XP_015312709.1 | 3 - 4 | 1383 | 93 | ||
XM_015457224.1 | XP_015312710.1 | 2 - 3 | 670 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
