Assay Details
Gene Symbol: LOC102133268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591430.1 | XP_005591487.1 | 23 - 24 | 3889 | 115 | |
XM_005591431.2 | XP_005591488.1 | 23 - 24 | 4298 | 115 | ||
GenBank mRNA | AB070033.1 | - | 4 - 5 | 731 | 115 | |
AB070190.1 | - | 6 - 7 | 707 | 115 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
