Assay Details
Gene Symbol: LOC102133268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591430.1 | XP_005591487.1 | 29 - 30 | 4931 | 140 | |
XM_005591431.2 | XP_005591488.1 | 29 - 30 | 5340 | 140 | ||
GenBank mRNA | AB070033.1 | - | 10 - 11 | 1773 | 140 | |
AB070190.1 | - | 12 - 13 | 1749 | 140 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
