Assay Details
Gene Symbol: LOC102133268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591430.1 | XP_005591487.1 | 3 - 4 | 422 | 122 | |
XM_005591431.2 | XP_005591488.1 | 3 - 4 | 831 | 122 | ||
GenBank mRNA | AB071112.1 | - | 3 - 4 | 393 | 122 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
