Assay Details
Gene Symbol: FAM47C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593266.2 | XP_005593323.1 | 1 | 3607 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM47C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593266.2 | XP_005593323.1 | 1 | 3607 | 88 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |