Assay Details
Gene Symbol: RHOXF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594475.2 | XP_005594532.1 | 4 - 5 | 791 | 68 | |
XM_005594476.2 | XP_005594533.1 | 4 - 5 | 791 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
