Assay Details
Gene Symbol: LOC102132156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553435.2 | XP_005553492.1 | 14 - 15 | 1947 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
