Assay Details
Gene Symbol: LOC102132156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553435.2 | XP_005553492.1 | 3 - 4 | 547 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
