Assay Details
| Gene Symbol: CD180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557059.2 | XP_005557116.1 | 2 - 3 | 421 | 109 | |
| XM_005557060.2 | XP_005557117.1 | 1 - 2 | 444 | 109 | ||
| XM_005557061.2 | XP_005557118.1 | 2 - 3 | 1704 | 109 | ||
| XM_015451578.1 | XP_015307064.1 | 3 - 4 | 505 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005557060.2, XM_005557061.2 transcripts.
Set Membership: |
|

CD180
Genomic Map