Assay Details
| Gene Symbol: LOC102142032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456085.1 | XP_015311571.1 | 5 - 6 | 2184 | 99 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102142032| Gene Symbol: LOC102142032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456085.1 | XP_015311571.1 | 5 - 6 | 2184 | 99 | |