Assay Details
| Gene Symbol: IL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015433396.1 | XP_015288882.1 | 4 - 5 | 952 | 90 | |
| XM_015433397.1 | XP_015288883.1 | 5 - 6 | 778 | 90 | ||
More Information
Set Membership: |
|

IL37
Genomic Map