Assay Details
| Gene Symbol: IL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015433396.1 | XP_015288882.1 | 6 - 7 | 1131 | 94 | |
| XM_015433397.1 | XP_015288883.1 | 7 - 8 | 957 | 94 | ||
More Information
Set Membership: |
|

IL37
Genomic Map