Assay Details
| Gene Symbol: ZNF195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576932.2 | XP_005576989.1 | 5 - 6 | 588 | 66 | |
| XM_005576933.2 | XP_005576990.1 | 4 - 5 | 489 | 66 | ||
| XM_005576934.2 | XP_005576991.1 | 4 - 5 | 489 | 66 | ||
| XM_015434302.1 | XP_015289788.1 | 3 - 4 | 409 | 66 | ||
| XM_015434303.1 | XP_015289789.1 | 5 - 6 | 588 | 66 | ||
| XM_015434308.1 | XP_015289794.1 | 5 - 6 | 589 | 66 | ||
More Information
Set Membership: |
|

ZNF195
Genomic Map