Assay Details
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581219.2 | XP_005581276.1 | 4 - 5 | 1234 | 74 | |
XM_015436952.1 | XP_015292438.1 | 4 - 5 | 908 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
