Assay Details
Gene Symbol: LOC102130441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549132.2 | XP_005549189.1 | 2 - 3 | 716 | 67 | |
XM_005549133.2 | XP_005549190.1 | 2 - 3 | 637 | 67 | ||
XM_005549134.2 | XP_005549191.1 | 3 - 4 | 646 | 67 | ||
XM_015447095.1 | XP_015302581.1 | 2 - 3 | 506 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
