Assay Details
| Gene Symbol: LOC102130441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549132.2 | XP_005549189.1 | 4 - 5 | 1067 | 73 | |
| XM_005549133.2 | XP_005549190.1 | 4 - 5 | 988 | 73 | ||
| XM_005549134.2 | XP_005549191.1 | 5 - 6 | 997 | 73 | ||
| XM_015447095.1 | XP_015302581.1 | 4 - 5 | 857 | 73 | ||
More Information
Set Membership: |
|

LOC102130441
Genomic Map