Assay Details
| Gene Symbol: LOC102133652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552114.2 | XP_005552171.1 | 5 - 6 | 1040 | 110 | |
| XM_015448956.1 | XP_015304442.1 | 5 - 6 | 1040 | 110 | ||
| XM_015448957.1 | XP_015304443.1 | 4 - 5 | 822 | 110 | ||
| XM_015448958.1 | XP_015304444.1 | 4 - 5 | 718 | 110 | ||
More Information
Set Membership: |
|

LOC102133652
Genomic Map