Assay Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557129.2 | XP_005557186.1 | 2 - 3 | 1419 | 80 | |
XM_005557130.2 | XP_005557187.1 | 2 - 3 | 1419 | 80 | ||
XM_005557131.2 | XP_005557188.1 | 2 - 3 | 852 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
