Assay Details
Gene Symbol: LOC102135463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015452260.1 | XP_015307746.1 | 2 - 3 | 215 | 76 | |
XM_015452261.1 | XP_015307747.1 | 2 - 3 | 195 | 76 | ||
XM_015452263.1 | XP_015307749.1 | 2 - 3 | 406 | 76 | ||
XM_015452264.1 | XP_015307750.1 | 2 - 3 | 1238 | 76 | ||
XM_015452265.1 | XP_015307751.1 | 2 - 3 | 188 | 76 | ||
XM_015452266.1 | XP_015307752.1 | 2 - 3 | 215 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015452260.1, XM_015452261.1, XM_015452263.1, XM_015452264.1, XM_015452265.1, XM_015452266.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
