Assay Details
Gene Symbol: LOC102135463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015452266.1 | XP_015307752.1 | 3 - 4 | 395 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015452266.1 transcripts.
Set Membership: |
![]() ![]() |
