Assay Details
| Gene Symbol: DUOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559422.2 | XP_005559479.1 | 2 - 3 | 1020 | 60 | |
| XM_015452782.1 | XP_015308268.1 | 1 - 2 | 1623 | 60 | ||
More Information
Set Membership: |
|

DUOXA1
Genomic Map