Assay Details
Gene Symbol: POLR1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589587.2 | XP_005589644.1 | 2 - 3 | 733 | 59 | |
XM_005589588.2 | XP_005589645.1 | 2 - 3 | 727 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
