Assay Details
| Gene Symbol: ZNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441012.1 | XP_015296498.1 | 2 - 3 | 126 | 75 | |
| XM_015441013.1 | XP_015296499.1 | 2 - 3 | 96 | 75 | ||
| XM_015441017.1 | XP_015296503.1 | 2 - 3 | 131 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015441012.1, XM_015441013.1, XM_015441017.1 transcripts.
Set Membership: |
|

ZNF135
Genomic Map