Assay Details
Gene Symbol: DCAF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015450206.1 | XP_015305692.1 | 3 | 2271 | 87 | |
XM_015450207.1 | XP_015305693.1 | 4 | 1606 | 87 | ||
XM_015450208.1 | XP_015305694.1 | 3 | 2090 | 87 | ||
XM_015450209.1 | XP_015305695.1 | 4 | 1415 | 87 | ||
XM_015450210.1 | XP_015305696.1 | 3 | 1322 | 87 | ||
XM_015450211.1 | XP_015305697.1 | 4 | 1410 | 87 | ||
XM_015450212.1 | XP_015305698.1 | 3 | 1307 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
